(5S)-5-(Trifluoromethyl)pyrrolidin-2-one ≥97%, ee 98%
Supplier: Apollo Scientific
PC520770-10G PC520770-1G PC520770-5G
APOSPC520770-10GEA 850 EUR
APOSPC520770-10G APOSPC520770-1G APOSPC520770-5G
(5S)-5-(Trifluoromethyl)pyrrolidin-2-one ≥97%, ee 98%
(5S)-5-(Trifluoromethyl)pyrrolidin-2-one
Formula: C₅H₆F₃NO
MW: 153.102 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22566326
CAS Number: 1287211-10-2