3,3-Difluoropiperidine-1-sulphonyl chloride 95%
Supplier: Apollo Scientific
PC520786-250MG PC520786-500MG
APOSPC520786-250MGEA 792 EUR
APOSPC520786-250MG APOSPC520786-500MG
3,3-Difluoropiperidine-1-sulphonyl chloride 95%
3,3-Difluoropiperidine-1-sulphonyl chloride
Formula: C₅H₈ClF₂NO₂S
MW: 219.637 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29074690
CAS Number: 1845716-99-5