1-Bromo-4-(difluoromethyl)-2-fluorobenzene 97%
Supplier: Apollo Scientific
PC520804-10G PC520804-1G PC520804-5G
APOSPC520804-10GEA 812 EUR
APOSPC520804-10G APOSPC520804-1G APOSPC520804-5G
1-Bromo-4-(difluoromethyl)-2-fluorobenzene 97%
1-Bromo-4-(difluoromethyl)-2-fluorobenzene
Formula: C₇H₄BrF₃
MW: 225.006 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698535
CAS Number: 1214386-70-5