5-Methyl-3-(trifluoromethyl)-1H-pyrazol-4-amine
Supplier: Apollo Scientific
PC520805-1G PC520805-250MG PC520805-500MG
APOSPC520805-1GEA 373 EUR
APOSPC520805-1G APOSPC520805-250MG APOSPC520805-500MG
5-Methyl-3-(trifluoromethyl)-1H-pyrazol-4-amine
5-Methyl-3-(trifluoromethyl)-1H-pyrazol-4-amine
Formula: C₅H₆F₃N₃
MW: 165.116 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04969915
CAS Number: 855343-08-7