1-(2,2,2-Trifluoroethyl)piperidin-4-amine hydrochloride 97%
Supplier: Apollo Scientific
PC520807-1G PC520807-250MG PC520807-500MG PC520807-100MG
APOSPC520807-1GEA 1440 EUR
APOSPC520807-1G APOSPC520807-250MG APOSPC520807-500MG APOSPC520807-100MG
1-(2,2,2-Trifluoroethyl)piperidin-4-amine hydrochloride 97%
1-(2,2,2-Trifluoroethyl)piperidin-4-amine hydrochloride
Formula: C₇H₁₃F₃N₂
MW: 182.187 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09040660
CAS Number: 187217-99-8