3-Fluoro-4-(4-methyl-1H-imidazol-1-yl)benzenamine 95%
Supplier: Apollo Scientific
PC520810-1G
PC520810-250MG
PC520810-500MG
APOSPC520810-1GEA
1290
EUR
APOSPC520810-1G
APOSPC520810-250MG
APOSPC520810-500MG
3-Fluoro-4-(4-methyl-1H-imidazol-1-yl)benzenamine 95%
3-Fluoro-4-(4-methyl-1H-imidazol-1-yl)benzenamine
Formula:
C₁₀H₁₀FN₃ MW: 191.205 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09834628 CAS Number: 252340-70-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...