2-Trifluoromethoxymethylpiperidine hydrochloride 97%
Supplier: Apollo Scientific
APOSPC520827-500MGEA 1350 EUR
APOSPC520827-500MG APOSPC520827-250MG
2-Trifluoromethoxymethylpiperidine hydrochloride 97%
2-Trifluoromethoxymethylpiperidine hydrochloride
Formula: C₇H₁₂F₃NO·ClH
MW: 219,632 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23381905
CAS Number: 2244088-33-1