1,1-Difluoro-5-azaspiro[2.3]hexane hydrochloride 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2,2-Difluoro-5-azaspiro[2.3]hexane hydrochloride
PC520858-250MG PC520858-100MG PC520858-1G
APOSPC520858-250MGEA 415 EUR
APOSPC520858-250MG APOSPC520858-100MG APOSPC520858-1G
1,1-Difluoro-5-azaspiro[2.3]hexane hydrochloride 97%
1,1-Difluoro-5-azaspiro[2.3]hexane hydrochloride
Formula: C₅H₇F₂N·ClH
MW: 155.574 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27987105
CAS Number: 1630906-91-0