2,4-Difluoro-5-methoxybenzotrifluoride
Supplier: Apollo Scientific
Warning
PC52119-1G PC52119-250MG
APOSPC52119-1GEA 406 EUR
APOSPC52119-1G APOSPC52119-250MG
2,4-Difluoro-5-methoxybenzotrifluoride
2,4-Difluoro-5-methoxybenzotrifluoride
Formula: C₈H₅F₅O
MW: 212.117 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28986197
CAS Number: 1935291-66-9