3-Bromo-5-(trifluoromethoxy)benzal fluoride
Supplier: Apollo Scientific
Warning
PC52181-1G PC52181-250MG PC52181-5G
APOSPC52181-1GEA 63.8 EUR
APOSPC52181-1G APOSPC52181-250MG APOSPC52181-5G
3-Bromo-5-(trifluoromethoxy)benzal fluoride
3-Bromo-5-(trifluoromethoxy)benzal fluoride
Formula: C₈H₄BrF₅O
MW: 291.013 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31812118
CAS Number: 2167884-91-3