Ethyl 2-hydroxy-4-methyl-2-(trifluoromethyl)pent-4-enoate 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-(Ethoxycarbonyl)-4-methyl-1,1,1-trifluoropent-4-en-2-ol
PC5263-1G PC5263-25G PC5263-5G
APOSPC5263-1GEA 21.9 EUR
APOSPC5263-1G APOSPC5263-25G APOSPC5263-5G
Ethyl 2-hydroxy-4-methyl-2-(trifluoromethyl)pent-4-enoate 97%
Ethyl 2-hydroxy-4-methyl-2-(trifluoromethyl)pent-4-enoate
Formula: C₉H₁₃F₃O₃
MW: 226.193 g/mol
Boiling Pt: 46 °C (2.3 mm Hg)
Density: 1.194 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD08532486
CAS Number: 217195-91-0