4-Bromo-2-chloro-3-fluorobenzoic acid 95%
Supplier: Apollo Scientific
PC52713-1G PC52713-250MG PC52713-5G
APOSPC52713-1GEA 527 EUR
APOSPC52713-1G APOSPC52713-250MG APOSPC52713-5G
4-Bromo-2-chloro-3-fluorobenzoic acid 95%
4-Bromo-2-chloro-3-fluorobenzoic acid
Formula: C₇H₃BrClFO₂
MW: 253.453 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28733905
CAS Number: 1807036-03-8