Methyl 4-bromo-2-chloro-3-fluorobenzoate 95%
Supplier: Apollo Scientific
PC52715-1G PC52715-250MG PC52715-5G
APOSPC52715-1GEA 387 EUR
APOSPC52715-1G APOSPC52715-250MG APOSPC52715-5G
Methyl 4-bromo-2-chloro-3-fluorobenzoate 95%
Methyl 4-bromo-2-chloro-3-fluorobenzoate
Formula: C₈H₅BrClFO₂
MW: 267.48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23707535
CAS Number: 1427390-96-2