3-[4-(Trifluoromethyl)phenoxy]azetidine hydrochloride 97%
Supplier: Apollo Scientific
PC530025-1G PC530025-250MG PC530025-500MG
APOSPC530025-1GEA 699 EUR
APOSPC530025-1G APOSPC530025-250MG APOSPC530025-500MG
3-[4-(Trifluoromethyl)phenoxy]azetidine hydrochloride 97%
3-[4-(Trifluoromethyl)phenoxy]azetidine hydrochloride
Formula: C₁₀H₁₀F₃NO·ClH
MW: 253.649 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18071794
CAS Number: 1236862-38-6