4-Fluoro-D-tryptophan 97%
Supplier: Apollo Scientific
PC530035-1G PC530035-250MG PC530035-500MG
APOSPC530035-1GEA 980 EUR
APOSPC530035-1G APOSPC530035-250MG APOSPC530035-500MG
4-Fluoro-D-tryptophan 97%
4-Fluoro-D-tryptophan
Formula: C₁₁H₁₁FN₂O₂
MW: 222.216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10565752
CAS Number: 110221-04-0