6-Fluoro-D-tryptophan 97%
Supplier: Apollo Scientific
PC530037-1G PC530037-250MG PC530037-500MG
APOSPC530037-1GEA 980 EUR
APOSPC530037-1G APOSPC530037-250MG APOSPC530037-500MG
6-Fluoro-D-tryptophan 97%
6-Fluoro-D-tryptophan
Formula: C₁₁H₁₁FN₂O₂
MW: 222.216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17214432
CAS Number: 108391-82-8