4-(Difluoromethoxy)-3-fluorobenzonitrile 95%
Supplier: Apollo Scientific
Warning
PC53061-1G PC53061-250MG PC53061-5G
APOSPC53061-1GEA 109 EUR
APOSPC53061-1G APOSPC53061-250MG APOSPC53061-5G
4-(Difluoromethoxy)-3-fluorobenzonitrile 95%
4-(Difluoromethoxy)-3-fluorobenzonitrile
Formula: C₈H₄F₃NO
MW: 187.119 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18394957
CAS Number: 954388-59-1
UN: 3276
ADR: 6.1 III