(2,6-Dibromo-4-fluorophenyl)methanol 97%
Supplier: Apollo Scientific
PC53089-100MG PC53089-1G PC53089-250MG
APOSPC53089-100MGEA 30.7 EUR
APOSPC53089-100MG APOSPC53089-1G APOSPC53089-250MG
(2,6-Dibromo-4-fluorophenyl)methanol 97%
(2,6-Dibromo-4-fluorophenyl)methanol
Formula: C₇H₅Br₂FO
Storage Temperature: Ambient
MDL Number: MFCD28717473
CAS Number: 1346674-69-8