3-(Trifluoromethoxy)propan-1-ol 97%
Supplier: Apollo Scientific
PC53096-1G PC53096-250MG PC53096-500MG
APOSPC53096-1GEA 931 EUR
APOSPC53096-1G APOSPC53096-250MG APOSPC53096-500MG
3-(Trifluoromethoxy)propan-1-ol 97%
3-(Trifluoromethoxy)propan-1-ol
Formula: C₄H₇F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD13186654
CAS Number: 949009-61-4