1H,1H,2H,2H-Perfluorotetradecan-1-ol 85%
Supplier: Apollo Scientific
Warning
PC53099-1G PC53099-250MG PC53099-5G
APOSPC53099-1GEA 422 EUR
APOSPC53099-1G APOSPC53099-250MG APOSPC53099-5G
1H,1H,2H,2H-Perfluorotetradecan-1-ol 85%
1H,1H,2H,2H-Perfluorotetradecan-1-ol
Formula: C₁₄H₅F₂₅O
MW: 664.149 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00792520
CAS Number: 39239-77-5