4-Fluoro-1-butanol
Supplier: Apollo Scientific
Warning
PC53179-10G PC53179-1G PC53179-5G
APOSPC53179-10GEA 134 EUR
APOSPC53179-10G APOSPC53179-1G APOSPC53179-5G
4-Fluoro-1-butanol
4-Fluoro-1-butanol
Formula: C₄H₉FO
MW: 92.112 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00039555
CAS Number: 372-93-0
UN: 1993
ADR: 3 III