2,6-Dichloro-4-fluorobenzaldehyde
Supplier: Apollo Scientific
Warning
PC53258-1G PC53258-5G PC53258-10G
APOSPC53258-1GEA 30.6 EUR
APOSPC53258-1G APOSPC53258-5G APOSPC53258-10G
2,6-Dichloro-4-fluorobenzaldehyde
2,6-Dichloro-4-fluorobenzaldehyde
Formula: C₇H₃Cl₂FO
MW: 193.003 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20526366
CAS Number: 1182709-86-9