Methyl 2-bromo-5-fluoro-3-nitrobenzoate
Supplier: Apollo Scientific
Warning
PC53305-1G PC53305-250MG PC53305-5G
APOSPC53305-1GEA 62.5 EUR
APOSPC53305-1G APOSPC53305-250MG APOSPC53305-5G
Methyl 2-bromo-5-fluoro-3-nitrobenzoate
Methyl 2-bromo-5-fluoro-3-nitrobenzoate
Formula: C₈H₅BrFNO₄
MW: 278.032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12172980
CAS Number: 328547-12-2