5-Fluoro-2-iodophenol
Supplier: Apollo Scientific
Warning
Synonyms: 5-Fluoro-2-iodo-phenol
PC53345-1G PC53345-250MG PC53345-5G
APOSPC53345-1GEA 306 EUR
APOSPC53345-1G APOSPC53345-250MG APOSPC53345-5G
5-Fluoro-2-iodophenol
5-Fluoro-2-iodophenol
Formula: C₆H₄FIO
MW: 237.998 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16999912
CAS Number: 186589-87-7