4-(1,1-Difluoroethyl)benzonitrile
Supplier: Apollo Scientific
Warning
PC53352-1G PC53352-250MG
APOSPC53352-1GEA 249 EUR
APOSPC53352-1G APOSPC53352-250MG
4-(1,1-Difluoroethyl)benzonitrile
4-(1,1-Difluoroethyl)benzonitrile
Formula: C₉H₇F₂N
MW: 167.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17015609
CAS Number: 55805-05-5