4-Chloro-5-fluoro-6-methoxy-2-methylpyrimidine 95+%
Supplier: Apollo Scientific
Warning
PC535039-100MG PC535039-250MG
APOSPC535039-100MGEA 56.7 EUR
APOSPC535039-100MG APOSPC535039-250MG
4-Chloro-5-fluoro-6-methoxy-2-methylpyrimidine 95+%
4-Chloro-5-fluoro-6-methoxy-2-methylpyrimidine
Formula: C₆H₆ClFN₂O
MW: 176.576 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23163182
CAS Number: 1383976-40-6