4-Bromo-7-fluoroindolin-2-one 97%
Supplier: Apollo Scientific
Warning
PC535077-1G PC535077-250MG
APOSPC535077-1GEA 134 EUR
APOSPC535077-1G APOSPC535077-250MG
4-Bromo-7-fluoroindolin-2-one 97%
4-Bromo-7-fluoroindolin-2-one
Formula: C₈H₅BrFNO
MW: 230.034 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15526615
CAS Number: 1260903-30-7