6-Bromo-2-(trifluoromethyl)oxazolo[5,4-b]pyridine 95%
Supplier: Apollo Scientific
Warning
PC535091-1G PC535091-250MG
APOSPC535091-1GEA 172 EUR
APOSPC535091-1G APOSPC535091-250MG
6-Bromo-2-(trifluoromethyl)oxazolo[5,4-b]pyridine 95%
6-Bromo-2-(trifluoromethyl)oxazolo[5,4-b]pyridine
Formula: C₇H₂BrF₃N₂O
MW: 267.003 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07375029
CAS Number: 886373-24-6