Methyl 2-fluoro-4-hydroxy-5-nitrobenzoate 98%
Supplier: Apollo Scientific
PC53513-1G PC53513-250MG PC53513-5G
APOSPC53513-1GEA 277 EUR
APOSPC53513-1G APOSPC53513-250MG APOSPC53513-5G
Methyl 2-fluoro-4-hydroxy-5-nitrobenzoate 98%
Methyl 2-fluoro-4-hydroxy-5-nitrobenzoate
Formula: C₈H₆FNO₅
MW: 215.135 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30534485
CAS Number: 2090453-52-2