3-[(2,2,3,3,4,4,5,5-Octafluoropent-1-yl)oxy]-1,2-propenoxide 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Glycidyl 2,2,3,3,4,4,5,5-octafluoropentyl ether, 2-(4,4,5,5,6,6,7,7-Octafluoro-2-oxaheptyl)oxirane, 2-{[(2,2,3,3,4,4,5,5-Octafluoropent-1-yl)oxy]methyl}oxirane
PC5353D-25G
PC5353D-5G
PC5353D-1G
APOSPC5353D-25GEA
439
EUR
APOSPC5353D-25G
APOSPC5353D-5G
APOSPC5353D-1G
3-[(2,2,3,3,4,4,5,5-Octafluoropent-1-yl)oxy]-1,2-propenoxide 97%
3-[(2,2,3,3,4,4,5,5-Octafluoropent-1-yl)oxy]-1,2-propenoxide
Formula:
C₈H₈F₈O₂ MW: 288.135 g/mol Boiling Pt: 75…79 °C (4 mm Hg) Density: 1.509 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00054692 CAS Number: 19932-27-5 |