2,6-Difluoro-3-(trifluoromethyl)phenol
Supplier: Apollo Scientific
Danger
PC53552-1G PC53552-250MG PC53552-5G
APOSPC53552-1GEA 1570 EUR
APOSPC53552-1G APOSPC53552-250MG APOSPC53552-5G
2,6-Difluoro-3-(trifluoromethyl)phenol
2,6-Difluoro-3-(trifluoromethyl)phenol
Formula: C₇H₃F₅O
Storage Temperature: Ambient
MDL Number: MFCD30537676
CAS Number: 2149589-77-3
UN: 1760
ADR: 8,III