1-(2-Fluoro-4-(trifluoromethyl)phenyl)cyclopropanecarboxylic acid 98%
Supplier: Apollo Scientific
Warning
PC56038-10G PC56038-1G PC56038-5G
APOSPC56038-10GEA 1180 EUR
APOSPC56038-10G APOSPC56038-1G APOSPC56038-5G
1-(2-Fluoro-4-(trifluoromethyl)phenyl)cyclopropanecarboxylic acid 98%
1-(2-Fluoro-4-(trifluoromethyl)phenyl)cyclopropanecarboxylic acid
Formula: C₁₁H₈F₄O₂
Storage Temperature: Ambient
MDL Number: MFCD19693145
CAS Number: 1274902-10-1