2,3-Difluoro-4-((trimethylsilyl)ethynyl)aniline
Supplier: Apollo Scientific
Warning
PC56087-1G PC56087-250MG PC56087-5G
APOSPC56087-1GEA 1320 EUR
APOSPC56087-1G APOSPC56087-250MG APOSPC56087-5G
2,3-Difluoro-4-((trimethylsilyl)ethynyl)aniline
2,3-Difluoro-4-((trimethylsilyl)ethynyl)aniline
Formula: C₁₁H₁₃F₂NSi
MW: 225.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30830087
CAS Number: 2149597-51-1