trans-4,5-Difluoro-1,3-dioxolan-2-one
Supplier: Apollo Scientific
Warning
PC56110-1G PC56110-250MG PC56110-5G
APOSPC56110-1GEA 405 EUR
APOSPC56110-1G APOSPC56110-250MG APOSPC56110-5G
trans-4,5-Difluoro-1,3-dioxolan-2-one
trans-4,5-Difluoro-1,3-dioxolan-2-one
Formula: C₃H₂F₂O₃
MW: 124.043 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31536706
CAS Number: 311810-76-1