1-(Perfluorobutyl)icosane 97%
Supplier: Apollo Scientific
Warning
PC56216-1G PC56216-250MG
APOSPC56216-1GEA 102 EUR
APOSPC56216-1G APOSPC56216-250MG
1-(Perfluorobutyl)icosane 97%
1-(Perfluorobutyl)icosane
Formula: C₂₄H₄₁F₉
MW: 500.568 g/mol
Melting Pt: 38…39 °C
Storage Temperature: Ambient
MDL Number: MFCD28135957
CAS Number: 808758-67-0