7-Fluoro-6-iodo-1H-indazole 95+%
Supplier: Apollo Scientific
Warning
PC56239-100MG PC56239-250MG
APOSPC56239-100MGEA 279 EUR
APOSPC56239-100MG APOSPC56239-250MG
7-Fluoro-6-iodo-1H-indazole 95+%
7-Fluoro-6-iodo-1H-indazole
Formula: C₇H₄FIN₂
MW: 262.023 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31580164
CAS Number: 2227272-43-5