2-Bromo-4,5-difluoroiodobenzene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Bromo-4,5-difluoro-2-iodobenzene
PC56258-1G PC56258-250MG PC56258-5G
APOSPC56258-1GEA 192 EUR
APOSPC56258-1G APOSPC56258-250MG APOSPC56258-5G
2-Bromo-4,5-difluoroiodobenzene 98%
2-Bromo-4,5-difluoroiodobenzene
Formula: C₆H₂BrF₂I
MW: 318.885 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20233325
CAS Number: 1349718-22-4