1,1-Difluoro-2-methylpropan-2-ol 95%
Supplier: Apollo Scientific
|
Danger
|
PC56324-1G
PC56324-250MG
APOSPC56324-1GEA
655
EUR
APOSPC56324-1G
APOSPC56324-250MG
1,1-Difluoro-2-methylpropan-2-ol 95%
1,1-Difluoro-2-methylpropan-2-ol
Formula:
C₄H₈F₂O MW: 110.103 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23943409 CAS Number: 354-10-9 UN: 1987 ADR: 3 III |