2-(2,6-Difluorophenyl)propionic acid 95%
Supplier: Apollo Scientific
Warning
PC56800-1G PC56800-250MG
APOSPC56800-1GEA 1010 EUR
APOSPC56800-1G APOSPC56800-250MG
2-(2,6-Difluorophenyl)propionic acid 95%
2-(2,6-Difluorophenyl)propionic acid
Formula: C₉H₈F₂O₂
MW: 186.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11036843
CAS Number: 359828-68-5