4-Bromo-2-fluoro-5-methylphenol
Supplier: Apollo Scientific
Warning
PC56835-1G PC56835-500MG PC56835-5G
APOSPC56835-1GEA 232 EUR
APOSPC56835-1G APOSPC56835-500MG APOSPC56835-5G
4-Bromo-2-fluoro-5-methylphenol
4-Bromo-2-fluoro-5-methylphenol
Formula: C₇H₆BrFO
MW: 205.024 g/mol
Melting Pt: 60…62 °C
Storage Temperature: Ambient
MDL Number: MFCD09261252
CAS Number: 550400-07-2