8-Bromo-6-(trifluoromethyl)imidazo[1,5-a]pyrimidine 95%
Supplier: Apollo Scientific
Danger
PC56877-1G PC56877-250MG
APOSPC56877-1GEA 654 EUR
APOSPC56877-1G APOSPC56877-250MG
8-Bromo-6-(trifluoromethyl)imidazo[1,5-a]pyrimidine 95%
8-Bromo-6-(trifluoromethyl)imidazo[1,5-a]pyrimidine
Formula: C₇H₃BrF₃N₃
Storage Temperature: Ambient
MDL Number: MFCD31664912
CAS Number: 2151834-34-1
UN: 2811
ADR: 6.1,III