5-Bromo-7-fluoro-benzofuran-2-carboxylic acid
Supplier: Apollo Scientific
Warning
PC56919-1G PC56919-250MG
APOSPC56919-1GEA 464 EUR
APOSPC56919-1G APOSPC56919-250MG
5-Bromo-7-fluoro-benzofuran-2-carboxylic acid
5-Bromo-7-fluoro-benzofuran-2-carboxylic acid
Formula: C₉H₄BrFO₃
MW: 259.029 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16743019
CAS Number: 1249506-21-5