2-Chloro-3,5-difluorobenzaldehyde 95%
Supplier: Apollo Scientific
Warning
PC56948-1G PC56948-250MG
APOSPC56948-1GEA 549 EUR
APOSPC56948-1G APOSPC56948-250MG
2-Chloro-3,5-difluorobenzaldehyde 95%
2-Chloro-3,5-difluorobenzaldehyde
Formula: C₇H₃ClF₂O
MW: 176.548 g/mol
Melting Pt: 59…60 °C
Storage Temperature: Ambient
MDL Number: MFCD18415552
CAS Number: 1261561-82-3