1-Bromo-3-(difluoromethyl)-5-fluorobenzene
Supplier: Apollo Scientific
Warning
PC56966-1G PC56966-250MG PC56966-5G
APOSPC56966-1GEA 60.9 EUR
APOSPC56966-1G APOSPC56966-250MG APOSPC56966-5G
1-Bromo-3-(difluoromethyl)-5-fluorobenzene
1-Bromo-3-(difluoromethyl)-5-fluorobenzene
Formula: C₇H₄BrF₃
MW: 225.006 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09261145
CAS Number: 627526-90-3