Methyl 2,2,3,3-tetrafluoropropionate
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Methyl 3H-tetrafluoropropanoate, Methyl 2,2,3,3-tetrafluoropropanoate
PC57058-25G
PC57058-5G
APOSPC57058-25GEA
865
EUR
APOSPC57058-25G
APOSPC57058-5G
Methyl 2,2,3,3-tetrafluoropropionate
Methyl 2,2,3,3-tetrafluoropropionate
Formula:
C₄H₄F₄O₂ MW: 160.067 g/mol Boiling Pt: 93 °C (774 mm Hg) Density: 1.3819 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD03412257 CAS Number: 1893-38-5 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...