4’-Chloro-2’-hydroxy-2,2,2-trifluoroacetophenone 95%
Supplier: Apollo Scientific
Warning
PC57071-1G PC57071-250MG PC57071-5G
APOSPC57071-1GEA 215 EUR
APOSPC57071-1G APOSPC57071-250MG APOSPC57071-5G
4’-Chloro-2’-hydroxy-2,2,2-trifluoroacetophenone 95%
4’-Chloro-2’-hydroxy-2,2,2-trifluoroacetophenone
Formula: C₈H₄ClF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD31922518
CAS Number: 2229260-52-8