2-Bromo-N-(2,2,2-trifluoroethyl)aniline
Supplier: Apollo Scientific
Warning
PC57086-1G PC57086-250MG
APOSPC57086-1GEA 42.2 EUR
APOSPC57086-1G APOSPC57086-250MG
2-Bromo-N-(2,2,2-trifluoroethyl)aniline
2-Bromo-N-(2,2,2-trifluoroethyl)aniline
Formula: C₈H₇BrF₃N
MW: 254.047 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11153061
CAS Number: 1021056-39-2