2-Acetamido-4-(trifluoromethyl)benzeneboronic acid, pinacol ester 98%
Supplier: Apollo Scientific
Synonyms: N-[2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)phenyl]acetamide, 3-(Acetylamino)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzotrifluoride, N-(2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)phenyl)acetamide
PC5715-100MG PC5715-500MG
APOSPC5715-100MGEA 52.6 EUR
APOSPC5715-100MG APOSPC5715-500MG
2-Acetamido-4-(trifluoromethyl)benzeneboronic acid, pinacol ester 98%
2-Acetamido-4-(trifluoromethyl)benzeneboronic acid, pinacol ester
Formula: C₁₅H₁₉BF₃NO₃
MW: 329.122 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12026072
CAS Number: 1150271-57-0