1-Bromo-3,3,4,4-tetrafluorohexane
Supplier: Apollo Scientific
Warning
PC5731-250MG PC5731-1G
APOSPC5731-250MGEA 284 EUR
APOSPC5731-250MG APOSPC5731-1G
1-Bromo-3,3,4,4-tetrafluorohexane
1-Bromo-3,3,4,4-tetrafluorohexane
Formula: C₆H₉BrF₄
MW: 237.033 g/mol
Boiling Pt: 93 °C (25 mm Hg)
Density: 1.46 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD12026453
CAS Number: 151831-48-0