Ethyl 3-oxo-3-(pentafluorophenyl)propanoate 97%
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl (pentafluorobenzoyl)acetate, Ethyl 3-oxo-3-(pentafluorophenyl)propionate
PC5767-1G PC5767-5G
APOSPC5767-1GEA 126 EUR
APOSPC5767-1G APOSPC5767-5G
Ethyl 3-oxo-3-(pentafluorophenyl)propanoate 97%
Ethyl 3-oxo-3-(pentafluorophenyl)propanoate
Formula: C₁₁H₇F₅O₃
Boiling Pt: 103…104 °C (4 mm Hg)
Density: 1.43 g/cm³ (25 °C)
Flash Pt: 41 °C
Storage Temperature: Ambient
MDL Number: MFCD00192163
CAS Number: 3516-87-8
UN: 3272
ADR: 3,III